Learn More
Sigma Aldrich Fine Chemicals Biosciences flt 3 Ligand human Animal
Catalog No. 502905027
Encompass
fLT3LG (fms-related tyrosine kinase 3 ligand) is a transmembrane glycoprotein and a ligand for fLT3 receptor. It is widely expressed in human tissues. The protein exists in membrane bound as well as secreted forms. Binding of fLT3LG to fLT3 results in receptor dimerization activation of the tyrosine kinase domain autophosphorylation and thereby activation of the receptor. Recombinant human flt3-Ligand is a soluble 17.6kDa protein consisting of 155 amino acid residues.
Catalog No. 50-290-5027 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP304410UG
fLT3LG (fms-related tyrosine kinase 3 ligand) is a transmembrane glycoprotein and a ligand for fLT3 receptor. It is widely expressed in human tissues. The protein exists in membrane bound as well as secreted forms. Binding of fLT3LG to fLT3 results in receptor dimerization activation of the tyrosine kinase domain autophosphorylation and thereby activation of the receptor. Recombinant human flt3-Ligand is a soluble 17.6kDa protein consisting of 155 amino acid residues.