Learn More
ACROBiosystems ACROBiosystems Mouse Flt-3 Ligand Protein, His Tag
Catalog No. 501957007
Encompass_Preferred
Recombinant Protein;100UG;Mouse Flt-3 Ligand, His Tag (FLL-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Arg 188 (Accession # P49772-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Flt-3, Fc Tag (Cat. No. FL3-H5258) at 5 μg/mL (100 μL/well) can bind Mouse Flt-3 Ligand, His Tag (Cat. No. FLL-M52H4) with a linear range of 0.1-0.6 ng/mL (QC tested).
Catalog No. 50-195-7007 Supplier ACROBiosystems Supplier No. FLLM52H4100UG
Recombinant Protein;100UG;Mouse Flt-3 Ligand, His Tag (FLL-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Arg 188 (Accession # P49772-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Flt-3, Fc Tag (Cat. No. FL3-H5258) at 5 μg/mL (100 μL/well) can bind Mouse Flt-3 Ligand, His Tag (Cat. No. FLL-M52H4) with a linear range of 0.1-0.6 ng/mL (QC tested).