Learn More
Eurofins Discoverx Flt-4, active, 250 µg. (14-681M)
Supplier:  Eurofins Discoverx 14681M
Encompass
N-terminal GST-tagged recombinant, human Flt4 amino acids 800-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9180