Learn More
Ankom Technology Fiber Filter Bags -100 count
SDP
Supplier:  Ankom Technology F58
Encompass
The F58 filter bag is mainly designed for customers working with small grind size samples (0.5 – 1.0 mm screens) to support NIR analysis and some samples known for significant pulverization during grinding, regardless of screen size. The porosity of the F58 filter bags are finer than the F57 but have increased flow capacity because of the type of polymer used to make the filter matrix. The F58 will allow better flow of chemicals to the sample and reduce artificially high values due to clumping caused by some sample types. Use for crude fiber, neutral detergent fiber and acid detergent fiber analysis. The bag has not been validated for use in the Daisy Incubator for in vitro studies. Due to the polymers used, the F58 bag cannot be used to support follow-on assays for nitrogen or 72% H2SO4 lignin.
Catalog No. NC1691343