Learn More
Neta Scientific Agilent Vial,HS,crimp,FB,10ml,clr,cert,100PK
SDP
Catalog No. 50929311
Encompass_Preferred
This item is manufactured by Agilent
Agilent Vial, crimp top, headspace, clear, certified, flat bottom, 10 mL, 23 x 46 mm, 100/pk. Vial size: 22.75 x 46 mm (20 mm cap) - Headspace Vials & Caps
UNSPSC Code  41121806
- 58
Catalog No. 50-929-311 Supplier Neta Scientific Supplier No. 51820838
This item is manufactured by Agilent
Agilent Vial, crimp top, headspace, clear, certified, flat bottom, 10 mL, 23 x 46 mm, 100/pk. Vial size: 22.75 x 46 mm (20 mm cap) - Headspace Vials & Caps
UNSPSC Code  41121806
- 58