Learn More
Neta Scientific Agilent Vial,HS,crimp,FB,20ml,clr,cert,100PK
SDP
Catalog No. 50929310
Encompass_Preferred
This item is manufactured by Agilent
Agilent Vial, headspace, certified, crimp, clear, flat bottom, 20 mL, 100/pk. Vial size: 22.75 x 75 mm (20 mm cap) - Headspace Vials & Caps
UNSPSC Code  41121806
- 58
Catalog No. 50-929-310 Supplier Neta Scientific Supplier No. 51820837
This item is manufactured by Agilent
Agilent Vial, headspace, certified, crimp, clear, flat bottom, 20 mL, 100/pk. Vial size: 22.75 x 75 mm (20 mm cap) - Headspace Vials & Caps
UNSPSC Code  41121806
- 58