Learn More
Alkali Scientific FLT, DSK, N66 5.0um Pore , 47mm Diameter, 100/pk
SDP
Catalog No. 5000662496
Encompass_Preferred
Disk filters made from high quality Nylon 66 membrane with a 5.0 µm pore and 47 mm diameter, designed for reliable particle retention and efficient filtration in laboratory workflows. These membrane filters provide excellent chemical compatibility, strong mechanical stability, and consistent flow rates, making them suitable for aqueous solutions, solvents, and general laboratory sample preparation. The uniform pore structure ensures dependable filtration performance and reproducible results in analytical, research, and quality control applications. Ideal for use in microbiology, environmental testing, and routine lab filtration procedures. Supplied as 100 filters per pack, these membranes support convenient handling and dependable filtration performance in professional laboratory environments.
Catalog No. 50-006-62496 Supplier Alkali Scientific Supplier No. XR812
Add to Cart
Add to Cart
Disk filters made from high quality Nylon 66 membrane with a 5.0 µm pore and 47 mm diameter, designed for reliable particle retention and efficient filtration in laboratory workflows. These membrane filters provide excellent chemical compatibility, strong mechanical stability, and consistent flow rates, making them suitable for aqueous solutions, solvents, and general laboratory sample preparation. The uniform pore structure ensures dependable filtration performance and reproducible results in analytical, research, and quality control applications. Ideal for use in microbiology, environmental testing, and routine lab filtration procedures. Supplied as 100 filters per pack, these membranes support convenient handling and dependable filtration performance in professional laboratory environments.