Learn More
MDI Membrane FLT MDI EHCN 47MM .45 EO 100PK
Catalog No. 501098767
Encompass_Preferred
mdi Cellulose Nitrate membrane filters with Hydrophobic Edge for Antibiotics, 47 mm, 0.45 µm, EO-sterile, 100 PK
Catalog No. 50-109-8767 Supplier MDI Membrane Supplier No. EHCN0902XXXX204
mdi Cellulose Nitrate membrane filters with Hydrophobic Edge for Antibiotics, 47 mm, 0.45 µm, EO-sterile, 100 PK