Learn More
MDI Membrane FLT MDI PTFE+GF 13MM .45 100PK
Supplier:  MDI Membrane SYTG0302MNXX104
Encompass_Preferred
mdi PTFE Syringe Filter with multi-layered Glass Fiber prefilter, 13 mm, 0.45 µm, Inlet/Outlet: Female luer lock/male luer slip, HPLC Certified, non-sterile, 100 PK
Catalog No. 50-109-8739