Learn More
MDI Membrane FLT PES 50MM 1/4INHB .2 STER 1
Supplier:  MDI Membrane VKSX1001BBXX302
Encompass_Preferred
mdi PES 50mm filter with vent (EFA-20 cm²), 0.45/0.2 µm, ¼" Hose Barb I/O, gamma sterilized, 10 PK
Catalog No. 50-109-8791