Learn More
MSC Ameripac Industries Bag, 16 x 48 in, 2 mil, 250/Case
Catalog No. 501916552
Encompass
Flat poly bags protect general light to medium weight products from dirt, dust, and moisture. These bags are manufactured from 100% virgin, high clarity polyethylene film and meet FDA and USDA specifications.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Suitable for protecting light to medium weight products
  • Can be closed with twist ties, bag tape, or impulse sealers
  • Ideal for general packaging and storage
Catalog No. 50-191-6552 Supplier MSC Supplier No. 39560941
Flat poly bags protect general light to medium weight products from dirt, dust, and moisture. These bags are manufactured from 100% virgin, high clarity polyethylene film and meet FDA and USDA specifications.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Suitable for protecting light to medium weight products
  • Can be closed with twist ties, bag tape, or impulse sealers
  • Ideal for general packaging and storage