Learn More
MSC Value Collection Polybags, 8 x 12 in, 2 mil, 1000/Pk
Catalog No. 501916834
Encompass
Polybags are versatile and flexible storage solutions, suitable for a wide range of applications from food to industrial tools and electronic equipment. These bags are manufactured from 100% virgin, high clarity polyethylene film and protect contents from dirt, dust, and moisture.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Can be closed with twist ties, bag tape, or an impulse sealer
  • Protects contents from dirt, dust, and moisture
  • Recyclable
  • Versatile and flexible storage solution
Catalog No. 50-191-6834 Supplier MSC Supplier No. 39563796
Polybags are versatile and flexible storage solutions, suitable for a wide range of applications from food to industrial tools and electronic equipment. These bags are manufactured from 100% virgin, high clarity polyethylene film and protect contents from dirt, dust, and moisture.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Can be closed with twist ties, bag tape, or an impulse sealer
  • Protects contents from dirt, dust, and moisture
  • Recyclable
  • Versatile and flexible storage solution