Learn More
Eurofins Discoverx Flt1, active, 250 µg. (14-923M)
Supplier:  Eurofins Discoverx 14923M
Encompass
N-terminal GST-tagged recombinant, human Flt1 amino acids 783-end. This protein encodes a member of the vascular endothelial growth factor receptor (VEGFR) family. VEGFR family members are receptor tyrosine kinases (RTKs) which contain an extracellular ligand-binding region with seven immunoglobulin (Ig)-like domains, a transmembrane segment, and a tyrosine kinase (TK) domain within the cytoplasmic domain. This protein binds to VEGFR-A, VEGFR-B and placental growth factor and plays an important role in angiogenesis and vasculogenesis. Expression of this receptor is found in vascular endothelial cells, placental trophoblast cells and peripheral blood monocytes. Multiple transcript variants encoding different isoforms have been found for this gene. Isoforms include a full length transmembrane receptor isoform and shortened, soluble isoforms. The soluble isoforms are associated with the onset of pre-eclampsia. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9420