Learn More
Sino Biological Recombinant Human FGFR3 Protein (d613-653, GST Tag)
Catalog No. 502574903
Encompass_Preferred
Recombinant human FGFR3 (d613-653) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4903 Supplier Sino Biological Supplier No. F0616HG100
Recombinant human FGFR3 (d613-653) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.