Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FLT3-IN-19 | 2237234-48-7 | C₂₂H₂₆N₈O | 5 MG
SDP

Catalog No. 5000373722
Encompass_Preferred

FLT3-IN-19 (Comp 50) is a potent and selective FLT3 inhibitor with an IC50 of 0.213 nM. It can be used in AML disease research.

  • Potent and selective FLT3 inhibitor (IC50 of 0.213 nM)
  • Suitable for AML disease research
  • Purity of 99.19%
  • Appearance as a solid
  • Color ranges from off-white to light yellow
  • Targets FLT3
  • Part of the protein tyrosine kinase/RTK pathway

Catalog No. 50-003-73722 Supplier Medchemexpress LLC Supplier No. HY1534735MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FLT3-IN-19 (Comp 50) is a potent and selective FLT3 inhibitor with an IC50 of 0.213 nM. It can be used in AML disease research.

  • Potent and selective FLT3 inhibitor (IC50 of 0.213 nM)
  • Suitable for AML disease research
  • Purity of 99.19%
  • Appearance as a solid
  • Color ranges from off-white to light yellow
  • Targets FLT3
  • Part of the protein tyrosine kinase/RTK pathway

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.