Learn More
Medchemexpress LLC FLT3 Mouse HEK293 10ug
SDP
Catalog No. 5000179446
Encompass_Preferred
FLT3 a receptor tyrosine kinase has diverse functions including kinase activation PI3K binding and ligase binding It plays a vital role in hematopoiesis immune function and tissue development FLT3 is expressed in various tissues like the brain immune system and reproductive system It is involved in diseases like leukemia and cancer The wide functions and tissue-specific expression emphasize FLT3 s importance in health and disease FLT3 Protein Mouse (HEK293 His) is the recombinant mouse-derived FLT3 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79446 Supplier Medchemexpress LLC Supplier No. HYP7415110UG
FLT3 a receptor tyrosine kinase has diverse functions including kinase activation PI3K binding and ligase binding It plays a vital role in hematopoiesis immune function and tissue development FLT3 is expressed in various tissues like the brain immune system and reproductive system It is involved in diseases like leukemia and cancer The wide functions and tissue-specific expression emphasize FLT3 s importance in health and disease FLT3 Protein Mouse (HEK293 His) is the recombinant mouse-derived FLT3 protein expressed by HEK293 with C-His labeled tag