Learn More
Sino Biological Recombinant Human FLT3 Protein (GST Tag), Active
Catalog No. 502574883
Encompass_Preferred
Recombinant human FLT3-ITD-W51 (571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4883 Supplier Sino Biological Supplier No. F1212CG50
Recombinant human FLT3-ITD-W51 (571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.