Learn More
Aviva Systems Biology Corp FLT3LG Antibody - N-terminal region (ARP45110_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45110_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLT3LG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FLT3LG. Alternate names: FL, FLT3LApplicationWB. Species reactivity: Cow, Dog, Human, Sheep
Catalog No. 50-014-58970