Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FLT3LG Human CHO 2ug
SDP

Catalog No. 5000196684
Encompass_Preferred

Recombinant human FLT3 ligand expressed in CHO cells and supplied as a lyophilized, glycosylated protein with confirmed biological activity in FLT3-responsive hematopoietic cell assays. Designed for research use in studies of hematopoiesis, progenitor cell proliferation, and FLT3 signaling.

  • Human species, CHO expression system
  • Lyophilized, glycosylated recombinant protein
  • Bioactivity ED50 < 1 ng/mL in cell proliferation assays
  • Endotoxin < 0.2 EU/μg (LAL)
  • Formulated with trehalose; reconstitute ≥100 μg/mL in ddH2O
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C for long term with carrier protein

Catalog No. 50-001-96684 Supplier Medchemexpress LLC Supplier No. HYP71112UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human FLT3 ligand expressed in CHO cells and supplied as a lyophilized, glycosylated protein with confirmed biological activity in FLT3-responsive hematopoietic cell assays. Designed for research use in studies of hematopoiesis, progenitor cell proliferation, and FLT3 signaling.

  • Human species, CHO expression system
  • Lyophilized, glycosylated recombinant protein
  • Bioactivity ED50 < 1 ng/mL in cell proliferation assays
  • Endotoxin < 0.2 EU/μg (LAL)
  • Formulated with trehalose; reconstitute ≥100 μg/mL in ddH2O
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C for long term with carrier protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.