Learn More
Medchemexpress LLC FLT3LG Mouse CHO 10ug
SDP
Supplier:  Medchemexpress LLC HYP735410UG
Encompass_Preferred
FLT3LG protein a potent stimulator of early hematopoietic cell proliferation activates FLT3 and synergizes with colony-stimulating factors and interleukins As a homodimer especially in isoform 2 it crucially promotes expansion and differentiation of hematopoietic progenitor cells FLT3LG s collaborative signaling with other molecules underscores its significance in regulating hematopoiesis and maintaining hematopoietic system balance FLT3LG Protein Mouse (CHO His) is the recombinant mouse-derived FLT3LG protein expressed by CHO cells with C-His labeled tag
Catalog No. 50-001-80063