Learn More
ACROBiosystems Recombinant Protein;Human Flt-3 Ligand Protein;InsectCells-HI5;100UG;FLL,FLT3LG,FL,FLT3L,FLT3-LG,FLT-3L,Flt3 ligand
Catalog No. 501657629
Encompass_Preferred
Recombinant Protein;Human Flt-3 Ligand Protein;100UG;Human Flt-3 Ligand (FLL-H55H7) is expressed from Baculovirus-Insect cells (HI5). It contains AA Thr 27 - Pro 185 (Accession # P49771-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.9 kDa. The protein migrates as 22-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7629 Supplier ACROBiosystems Supplier No. FLLH55H7100UG
Recombinant Protein;Human Flt-3 Ligand Protein;100UG;Human Flt-3 Ligand (FLL-H55H7) is expressed from Baculovirus-Insect cells (HI5). It contains AA Thr 27 - Pro 185 (Accession # P49771-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.9 kDa. The protein migrates as 22-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.