Learn More
Grainger Approved Vendor Threaded Rod, Steel, 5/8 in.-11, Grade A, 12 ft., 1 ea
Catalog No. 502783122
Encompass_Preferred
This threaded rod is a high-quality steel fastener designed for versatile industrial applications. It features a right-hand coarse thread and is manufactured with a rolled thread process to ensure structural integrity. This rod is finished with black oxide for improved performance and is suitable for environments within a temperature range of -58°F to +302°F.
  • Constructed from grade A steel for reliable strength
  • Features a 5/8 in. - 11 coarse thread size
  • Designed with right-hand thread direction and 1A thread fit
  • Provides a minimum yield strength of 45,000 psi and 60,000 psi tensile strength
  • Maintains a Rockwell hardness of B80 to B90
  • Meets RoHS and REACH compliance standards
Catalog No. 50-278-3122 Supplier Grainger Supplier No. 19NM49
This threaded rod is a high-quality steel fastener designed for versatile industrial applications. It features a right-hand coarse thread and is manufactured with a rolled thread process to ensure structural integrity. This rod is finished with black oxide for improved performance and is suitable for environments within a temperature range of -58°F to +302°F.
  • Constructed from grade A steel for reliable strength
  • Features a 5/8 in. - 11 coarse thread size
  • Designed with right-hand thread direction and 1A thread fit
  • Provides a minimum yield strength of 45,000 psi and 60,000 psi tensile strength
  • Maintains a Rockwell hardness of B80 to B90
  • Meets RoHS and REACH compliance standards