Learn More
Parker Hannifan FLTR AIR PTFE HF TTPOR 2 10
SDP
Catalog No. NC1917831
Encompass_Preferred
Parker domnick hunter HIGH FLOW TETPOR II Air/Gas Filters, Core Material: Stainless Steel, Endcap Configuration: C Style, Endcap Type: Locking 226, Length: 10 in., 25.4cm, Certifications/Compliance: Conforms to current USP Plastics Class VI - 121 deg.C, ASTM F838-83, Filter Efficiency: 0.2um
Catalog No. NC1917831 Supplier Parker Hannifan Supplier No. ZHFT/1C
Parker domnick hunter HIGH FLOW TETPOR II Air/Gas Filters, Core Material: Stainless Steel, Endcap Configuration: C Style, Endcap Type: Locking 226, Length: 10 in., 25.4cm, Certifications/Compliance: Conforms to current USP Plastics Class VI - 121 deg.C, ASTM F838-83, Filter Efficiency: 0.2um