Learn More
SKC Inc Opaque Solvent-resistant Polypropylene Filter Cassette Blank, 25 mm, 50 per pack
SDP
Catalog No. NC2867579
Encompass
Opaque Solvent-resistant White Polypropylene Filter Cassette Blank, 3 sections, 25 mm, goblet style with 1/2-inch spacer, leak-free, pk/50 DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC2867579 Supplier SKC Inc Supplier No. 2258585
Opaque Solvent-resistant White Polypropylene Filter Cassette Blank, 3 sections, 25 mm, goblet style with 1/2-inch spacer, leak-free, pk/50 DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”