Learn More
Parker Hannifan FLTR CPSL PEPLN PLS 0.6UM 3PK
SDP
Supplier:  Parker Hannifan FEPPB.60GGPN3
Encompass_Preferred
Parker domnick hunter Demicap Filter Capsules, Peplyn Plus Membrane - 5.5 in. (140mm) Length, Pore Size: 0.6um, Inlet Style: Stepped Hosebarb, Outlet Style: Stepped Hosebarb, Material: Polypropylene, Autoclavable: Yes
Catalog No. NC1917827