Learn More
Parker Hannifan FLTR CPSL PREPOR PES 3PK
SDP
Supplier:  Parker Hannifan FEPSB020HHPS3
Encompass_Preferred
Parker domnick hunter Demicap Filter Capsules, Prepor PES Membrane - 0.2um Pore Size, Filtration Area: 0.12m2, Inlet Style: 1/2 in. Hosebarb, Length: 140mm, 5.5 in., Sterile, Material: Polyethersulfone, Pore Size: 0.2um, Autoclavable: Yes
Catalog No. NC1917823