Learn More
Chemglass Life Sciences Filter Funnel, Disposable, 300mL, 20 Micron Polyethylene Frit
SDP
Catalog No. NC0755148
Encompass_Preferred
Filter Funnel, Disposable, 300mL, 40 Micron Polyethylene Frit, Case of 30
Catalog No. NC0755148 Supplier Chemglass Life Sciences Supplier No. OP660216
Filter Funnel, Disposable, 300mL, 40 Micron Polyethylene Frit, Case of 30