Learn More
MSC Made in USA, 16X25X2 1/8" 16GGE STL FLTR FRAME W/2CLIPS &SEAL
Catalog No. 5000509866
Encompass
This filter frame is designed for secure mounting and alignment of industrial filtration components. Constructed from 16-gauge steel, it provides structural integrity and durability for demanding applications.
  • Constructed from 16-gauge steel for enhanced strength
  • Includes two clips and a seal to ensure proper fitment and performance
  • Designed for 16 in. x 25 in. x 2.125 in. filtration systems
Catalog No. 50-005-09866 Supplier MSC Supplier No. 59828087
This filter frame is designed for secure mounting and alignment of industrial filtration components. Constructed from 16-gauge steel, it provides structural integrity and durability for demanding applications.
  • Constructed from 16-gauge steel for enhanced strength
  • Includes two clips and a seal to ensure proper fitment and performance
  • Designed for 16 in. x 25 in. x 2.125 in. filtration systems