Learn More
MSC Made In USA Filter Frame, 16 Gauge Steel, 12 in. x 24 in., 1 ea.
Catalog No. 5000509500
Encompass
This filter frame is designed for secure filter mounting and containment in industrial ventilation or filtration systems. It features a robust steel construction for durability and includes essential components for effective installation and sealing.
  • Constructed from 16-gauge steel
  • Dimensions are 12 in. x 24 in. x 2 1/8 in.
  • Includes two clips for secure attachment
  • Integrated seal design for improved performance
Catalog No. 50-005-09500 Supplier MSC Supplier No. 59828061
This filter frame is designed for secure filter mounting and containment in industrial ventilation or filtration systems. It features a robust steel construction for durability and includes essential components for effective installation and sealing.
  • Constructed from 16-gauge steel
  • Dimensions are 12 in. x 24 in. x 2 1/8 in.
  • Includes two clips for secure attachment
  • Integrated seal design for improved performance