Learn More
Chemglass Life Sciences 30mL Filter Funnel, Buchner, 14/20 Lower Vacuum Assembly, Coarse Frit
SDP
Catalog No. CG140648
Encompass_Preferred
30mL Filter Funnel, Buchner, 14/20 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.
Catalog No. CG140648 Supplier Chemglass Life Sciences Supplier No. CG140648
30mL Filter Funnel, Buchner, 14/20 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.