Learn More
Chemglass Life Sciences 15mL Filter Funnel, Buchner, 14/20 Lower Vacuum Assembly, Fine Frit
SDP
Catalog No. CG140640
Encompass_Preferred
15mL Filter Funnel, Buchner, 14/20 Lower Vacuum Assembly, Fine Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.
Catalog No. CG140640 Supplier Chemglass Life Sciences Supplier No. CG140640
Add to Cart
Add to Cart
15mL Filter Funnel, Buchner, 14/20 Lower Vacuum Assembly, Fine Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.