Learn More
Chemglass Life Sciences 15mL Filter Funnel, Buchner, 19/22 Lower Vacuum Assembly, Medium Frit
SDP
Catalog No. CG140644
Encompass_Preferred
15mL Filter Funnel, Buchner, 19/22 Lower Vacuum Assembly, Medium Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.
Catalog No. CG140644 Supplier Chemglass Life Sciences Supplier No. CG140644
15mL Filter Funnel, Buchner, 19/22 Lower Vacuum Assembly, Medium Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.