Learn More
Chemglass Life Sciences 30mL Filter Funnel, Buchner, Improved, 24/40 Lower Vacuum Assembly, Coarse Frit
SDP
Catalog No. CG1406E01
Encompass_Preferred
30mL Filter Funnel, Buchner, Improved, 24/40 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402. Improved design incorporates a delivery stem being sealed within the inner joint to prevent breakage. Funnel is supplied with a 3in piece of PTFE tubing that slides over the delivery stem. The tubing permits the funnel to be used during filtrations at higher vacuums without having the fluid pulled into the vacuum source. Overall height is without the PTFE tubing.
Catalog No. CG1406E01 Supplier Chemglass Life Sciences Supplier No. CG1406E01
30mL Filter Funnel, Buchner, Improved, 24/40 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402. Improved design incorporates a delivery stem being sealed within the inner joint to prevent breakage. Funnel is supplied with a 3in piece of PTFE tubing that slides over the delivery stem. The tubing permits the funnel to be used during filtrations at higher vacuums without having the fluid pulled into the vacuum source. Overall height is without the PTFE tubing.