Learn More
Chemglass Life Sciences 30mL Filter Funnel, Buchner, Coarse Frit
SDP
Catalog No. CG140210
Encompass_Preferred
30mL Filter Funnel, Buchner, Coarse Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.
Catalog No. CG140210 Supplier Chemglass Life Sciences Supplier No. CG140210
30mL Filter Funnel, Buchner, Coarse Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.