Learn More
Chemglass Life Sciences 600mL Filter Funnel, Buchner, Coarse Frit
SDP
Catalog No. CG140226
Encompass_Preferred
600mL Filter Funnel, Buchner, Coarse Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.
Catalog No. CG140226 Supplier Chemglass Life Sciences Supplier No. CG140226
Add to Cart
Add to Cart
600mL Filter Funnel, Buchner, Coarse Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.