Learn More
Chemglass Life Sciences 2mL Filter Funnel, Buchner, Fine Frit
SDP
Catalog No. CG140204
Encompass_Preferred
2mL Filter Funnel, Buchner, Fine Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.
Catalog No. CG140204 Supplier Chemglass Life Sciences Supplier No. CG140204
2mL Filter Funnel, Buchner, Fine Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.