Learn More
Chemglass Life Sciences 2L Filter Funnel, Buchner, Medium Frit
SDP
Catalog No. CG140231
Encompass_Preferred
2L Filter Funnel, Buchner, Medium Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.
Catalog No. CG140231 Supplier Chemglass Life Sciences Supplier No. CG140231
2L Filter Funnel, Buchner, Medium Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.