Learn More
Cytiva (Formerly Pall Lab) Acrodisc PSF Syringe Filters with PVDF Membrane - GxF/0.2 m, AutoPack tubes (25/pkg, 200/cs)
Supplier:  Cytiva AP4792
Encompass
Syringe filters with PVDF membrane are ideal for the filtration of many organic and non-organic solvents in HPLC (0.45 m) and UHPLC (0.2 m) sample preparation. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.PVDF syringe filters are chemical compatibility with aggressive acids and alcohols.Acrodisc PSF Syringe Filters have a large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL.Available with a built-in GxF prefilter for use with difficult-to-filter solutions.Acrodisc PSF Syringe Filters are designed to be fully compatible with automated robotic handling systems. The membrane and housing are extremely low in levels of potential extractables and are HPLC certified for low levels of UV-absorbing extractables. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3424