Learn More
MDI Membrane FLTR MDI CN 47MM 0.2 EO 100PK
Catalog No. 501098762
Encompass_Preferred
mdi Cellulose Nitrate membrane filters, 47 mm, 0.2 µm, EO-sterile, 100 PK
Catalog No. 50-109-8762 Supplier MDI Membrane Supplier No. CNXX0901XXXX204
mdi Cellulose Nitrate membrane filters, 47 mm, 0.2 µm, EO-sterile, 100 PK