Learn More
MDI Membrane FLTR MDI NYLN 13MM 0.45 100PK
Supplier:  MDI Membrane SYNN0302MNXX104
Encompass_Preferred
mdi Nylon-66 Syringe Filter, 13 mm, 0.45 µm, Inlet/Outlet: Female luer lock/male luer slip, HPLC Certified, non-sterile, 100 PK
Catalog No. 50-109-8713