Learn More
MDI Membrane FLTR MDI NYLN 47MM 0.45 100PK
Supplier:  MDI Membrane HNNX0902XXXX104
Encompass_Preferred
mdi Nylon-66 Disc filters, 47 mm, 0.45 µm, HPLC Certified, non-sterile, 100 PK
Catalog No. 50-109-8759