Learn More
MDI Membrane FLTR MDI PES 13MM .45 EO 100PK
Supplier:  MDI Membrane SYPL0302MNXX204
Encompass_Preferred
mdi PES Syringe filter, 13 mm, 0.45 µm, Inlet/Outlet: Female luer lock/male luer slip, ETO sterile, 100 PK
Catalog No. 50-109-8771