Learn More
MDI Membrane FLTR MDI PES 25MM 0.2 EO 100PK
Supplier:  MDI Membrane SYPL0601MNXX204
Encompass_Preferred
mdi PES Syringe filter, 25 mm, 0.2 µm, Inlet/Outlet: Female luer lock/male luer slip, ETO sterile, 100 PK
Catalog No. 50-109-8768