Learn More
MDI Membrane FLTR MDI PTFE 25MM 0.2 100PK
Supplier:  MDI Membrane SYTF0601MNXX104
Encompass_Preferred
mdi PTFE Syringe Filter, 25 mm, 0.2 µm, Inlet/Outlet: Female luer lock/male luer slip, HPLC Certified, non-sterile, 100 PK
Catalog No. 50-109-8732