Learn More
MDI Membrane FLTR MDI PTFE+GF 25MM .2 100PK
Supplier:  MDI Membrane SYTG0601MNXX104
Encompass_Preferred
mdi PTFE Syringe Filter with multi-layered Glass Fiber prefilter, 25 mm, 0.2 µm, Inlet/Outlet: Female luer lock/male luer slip, HPLC Certified, non-sterile, 100 PK
Catalog No. 50-109-8738