Learn More
IDEX Health & Science Upchurch FLTR UHMWPE SOLV CUPS 10U 5/PK
Catalog No. NC1920945
Encompass_Preferred
Idex Inlet Solvent Filters: Filter Cups, For Use With: Filters A-426/445/446/447, UHMWPE, Pore Size: 10 um, Replacement Filter Cup, Flow Rate: 5 mL/min
Catalog No. NC1920945 Supplier IDEX Health & Science Upchurch Supplier No. A427
Idex Inlet Solvent Filters: Filter Cups, For Use With: Filters A-426/445/446/447, UHMWPE, Pore Size: 10 um, Replacement Filter Cup, Flow Rate: 5 mL/min