Learn More
Health Care Logistics Inc Filtered Venting Needles, 20-gauge, 1-1/2", White, 25/pk
SDP
Catalog No. NC1553915
Encompass
Sterile filtered venting needle is designed for use by pharmacists and other specialists to evacuate drug containers during medication preparation and delivery. Can be used with any vial or bottle. 0.2 micron hydrophobic filter Yellow transfer needle is compact, easy-to-handle and cost effective compared to other venting needles. 20 gauge x 1½" stainless steel anti-coring needle easily penetrates all stoppers with minimum risk of damage. Maintains asepsis and prevents toxic, aerosolized materials from leaving the container during filling.
This item is not returnable. View return policy
Catalog No. NC1553915 Supplier Health Care Logistics Inc Supplier No. 18955
Sterile filtered venting needle is designed for use by pharmacists and other specialists to evacuate drug containers during medication preparation and delivery. Can be used with any vial or bottle. 0.2 micron hydrophobic filter Yellow transfer needle is compact, easy-to-handle and cost effective compared to other venting needles. 20 gauge x 1½" stainless steel anti-coring needle easily penetrates all stoppers with minimum risk of damage. Maintains asepsis and prevents toxic, aerosolized materials from leaving the container during filling.