Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics Flu A/Flu B/SARS-CoV-2 Control Panel
SDP

Catalog No. 21377164 Shop All Microbiologics Products
Change view
Click to view available options
Disposable:
Single-use
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
21-377-164 Single-use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-377-164 Supplier Microbiologics Supplier No. CL00016

Flu A/Flu B/SARS-CoV-2 Control Panel Helix Elite QC Sets help laboratories ensure the accuracy and reliability of their testing methods for detecting Influenza A (Flu A), Influenza B (Flu B), and SARS-CoV-2 viruses.

Contains:

  • 2 individually packaged posative control swabs
  • 2 individually packaged negative control swabs

Specifications

CE Marker No
Clia Complexity Not Applicable
Detectable Analytes Influenza A, Influenza B, SARS-CoV-2
Type Control Panel
Disposable Single-use
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.