Learn More
Microbiologics™ Flu/RSV/SARS-CoV-2 Control Panel (Inactivated Swab)
SDP
Catalog No. 23047383 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
12 Swabs
Catalog No. Quantity
23-047-383 12 Swabs
1 options
This item is not returnable. View return policy
Catalog No. 23-047-383 Supplier Microbiologics™ Supplier No. 8246
Add to Cart
Add to Cart
This item is not returnable. View return policy
Monitors the extraction, amplification and detection of Influenza A, Influenza B, Respiratory Syncytial Virus, and SARS-CoV-2 Virus molecular applications
  • Monitors the extraction, amplification and detection of Influenza A, Influenza B, Respiratory Syncytial Virus, and SARS-CoV-2 Virus molecular applications.
  • Routine use of external quality controls monitor day to day performance of molecular testing procedures.
Specifications
Biosafety Level 1
Sample Type Swab
Quantity 12 Swabs