Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NATtrol™ Flu/RSV/SARS-CoV-2 Positive Control Swab
SDP

Catalog No. 22158471 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
12/Pk
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-471 12/Pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-471 Supplier NATtrol™ Supplier No. NATFRCSW
Add to Cart
Add to Cart

Flu/RSV/SARS-CoV-2 Positive Control Swab (NATFRC-SW) is a qualitative, external control intended to evaluate the performance of molecular assays which detect Influenza A, Influenza B, RSV B and SARS-CoV-2 nucleic acids.

  • Contains purified, intact viral particles that have been chemically modified to render them non-infectious and human A-549 cells
  • Each swab contains Influenza A (A/Brisbane/10/07), Influenza B (B/Florida/02/06), RSV B (CH93(18)-18), SARS-CoV-2 (USA-WA1/2020)1 and A-549 cells
  • Analytes are formulated in a proprietary matrix and dried on swabs
  • Full process control designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures

Specifications

Manufacturing Origin United States
Product Type Positive Control
Sterility Non-sterile
Target Organism Class Influenza A Nucleic Acids, Influenza B Nucleic Acids, RSV-B Nucleic Acids, SARS-CoV-2 Nucleic Acids
Test Type Qualitative QC
Quantity 12/Pk
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.